missing translation for 'onlineSavingsMsg'
Learn More

Iron(II,III) oxide, 97% (metals basis)

Product Code. 11321769
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 11321769

Brand: Thermo Scientific Alfa Aesar 012374.C4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-61-9 | Fe3O4 | 231.53 g/mol

Mostly it is used as a black- pigment -catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning, whereas it is used as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymer, rubber, filler, building, construction, appliances, and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
Synonym Ferrosoferric oxide, Ferric ferrous oxide
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

Melting Point 1538°C
Physical Form Powder
Quantity 25 kg
Assay Percent Range 97% (metals basis)
Solubility Information Insoluble in water and organic solvents. Soluble in concentrated mineral acids.
Formula Weight 231.54
Refractive Index 3
Density 5.2 g/mL
Chemical Name or Material Iron(II, III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.