missing translation for 'onlineSavingsMsg'
Learn More

INO80 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 18810-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838654

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 15-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
Spezifikation

Spezifikation

INO80
Polyclonal
Unconjugated
INO80
hINO80, INO80, INO80 homolog (S. cerevisiae), INO80A, INOC1, KIAA1259
Rabbit
Antigen Affinity Chromatography
RUO
296084, 54617, 68142
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZPV2, Q9ULG1
INO80
Peptide
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote