missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL20RB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20521-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
IL20RB and IL20RA.[supplied by OMIM, Feb 2009].Spezifikation
IL20RB | |
Polyclonal | |
Unconjugated | |
IL20RB | |
DIRS1, FNDC6, IL 20 receptor subunit beta, IL 20R beta, IL 20R2, IL 20RB, IL20RB, interleukin 20 receptor beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53833 | |
-20°C | |
Liquid |
Immunocytochemistry, Flow Cytometry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXL0 | |
IL20RB | |
IL20RB Fusion Protein Ag14368 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |