Learn More
IL-6 Rat anti-Mouse, PE, Clone: MP5-20F3, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554401
Additional Details : Gewicht : 0.01000kg
Beschreibung
The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The immunogen used to generate the MP5-20F3 hybridoma was recombinant mouse IL-6. This is a neutralizing antibody.
Host Species: Rat
Clone: MP5-20F3
Isotype: IgG1
Species Reactivity: Mouse
Intracellular Staining
Spezifikation
IL-6 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-6 Recombinant Protein | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 |
Flow Cytometry | |
MP5-20F3 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.