missing translation for 'onlineSavingsMsg'
Learn More

IL-6 Rat anti-Human, PE, Clone: MQ2-6A3, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 554697

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827848

  • 630.00 CHF / 1 Stück
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody.

Host Species: Rat
Clone: MQ2-6A3
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant human IL-6

Intracellular Staining

Spezifikation

Spezifikation

IL-6
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Recombinant human IL-6
0.1 mg
Immunology
Human
IgG2a κ
Flow Cytometry
MQ2-6A3
PE
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.