missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Rat anti-Human, PE, Clone: MQ2-6A3, BD
Rat Monoclonal Antibody
630.00 CHF - 705.00 CHF
Spezifikation
Antigen | IL-6 |
---|---|
Klon | MQ2-6A3 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827848
|
BD Biosciences
554697 |
0.1 mg |
630.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15879708
|
BD Biosciences
559331 |
100 Tests |
705.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody.
Host Species: Rat
Clone: MQ2-6A3
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant human IL-6
Intracellular Staining
Spezifikation
IL-6 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
MQ2-6A3 | |
Flow Cytometry | |
PE | |
RUO | |
Human | |
Recombinant human IL-6 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.