Learn More
IL-5 Rat anti-Mouse/Human, APC, Clone: TRFK5, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554396
Additional Details : Gewicht : 0.01000kg
Beschreibung
The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and cross-reacts with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Host Species: Rat
Clone: TRFK5
Isotype: IgG1 κ
Species Reactivity: Mouse, Human
Immunogen: Mouse Semi-Purified T-Cell Clone Supernatant
Intracellular Staining
Spezifikation
IL-5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Semi-Purified T-Cell Clone Supernatant | |
0.1 mg | |
Immunology | |
Human, Mouse | |
IgG1 κ |
Flow Cytometry | |
TRFK5 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.