Learn More
IL-2 Rat anti-Mouse, FITC, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554427
Additional Details : Gewicht : 0.01000kg
Beschreibung
The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2). The immunogen used to generate the JES6-5H4 hybridoma was recombinant mouse IL-2. This is a neutralizing antibody.
Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Recombinant mouse IL-2
Intracellular Staining
Spezifikation
IL-2 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse IL-2 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b |
Flow Cytometry | |
JES6-5H4 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.