Learn More
IL-2 Rat anti-Mouse, FITC, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
158.00 CHF - 745.00 CHF
Spezifikation
Antigen | IL-2 |
---|---|
Klon | JES6-5H4 |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15831589
|
BD Biosciences
562040 |
25 μg |
158.00 CHF
1 Stück |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15887688
|
BD Biosciences
554427 |
0.1 mg |
745.00 CHF
1 Stück |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The JES6-5H4 monoclonal antibody specifically binds to mouse interleukin-2 (IL-2). IL-2 is a multifunctional cytokine that plays pivotal roles in immunity and tolerance. It is produced by activated T cells. IL-2 effects the activation, growth, proliferation and/or differentiation of various cell types including T and B lymphocytes and their precursors, LAK cells, NK cells, and monocytes/macrophages. IL-2 mediates its biological activities by binding to IL-2 receptor compleses. The intermediate affinity IL-2R is comprised of IL-2R β (CD122) and common gamma chain ( γc; CD132) subunits whereas the high-affinity IL-2R is comprised of IL-2R α (CD25), IL-2R β and γc subunits. The JES6-5H4 monoclonal antibody binds to IL-2 and neutralizes its biological activity.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
IL-2 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG2b | |
Affinity Purified |
JES6-5H4 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.