missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Mouse anti-Human, Mouse, Rat, Clone: 2C10B11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66526-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some maligcies. IMP-3 is expressed in maligt melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of maligt melanoma and may play an important role in the regulation of the biologic behavior of this tumor.Spezifikation
IGF2BP3 | |
Monoclonal | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00425, Q9CPN8 | |
Igf2bp3 | |
IGF2BP3 Fusion Protein Ag6349 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
2C10B11 | |
Unconjugated | |
IGF2BP3 | |
hKOC, IGF II mRNA binding protein 3, IGF2 mRNA binding protein 3, IGF2BP3, IMP 3, IMP3, KOC1, VICKZ family member 3, VICKZ3 | |
Mouse | |
Protein G | |
RUO | |
10643, 140488, 312320 | |
-20°C | |
Liquid |