missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNFN1A5 Partial ORF (NP_071911.2, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00064376-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16169626

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Members of the Ikaros (ZNFN1A1; MIM 603023) family of transcription factors, which includes Pegasus, are expressed in lymphocytes and are implicated in the control of lymphoid development.[supplied by OMIM]

Sequence: GEKKPEPLDFVKDFQEYLTQQTHHVNMISGSVSGDKEAEALQGAGTDGDQNGLDHPSVEVSLDENSGMLVDGFERTFDGKLKCRYCNYASKGTARLIEH
Spezifikation

Spezifikation

NP_071911.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GEKKPEPLDFVKDFQEYLTQQTHHVNMISGSVSGDKEAEALQGAGTDGDQNGLDHPSVEVSLDENSGMLVDGFERTFDGKLKCRYCNYASKGTARLIEH
RUO
IKZF5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
64376
ZNFN1A5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781B0249/FLJ22973/PEGASUS/ZNFN1A5
IKZF5
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote