Learn More
Abnova™ Human ZNFN1A5 Partial ORF (NP_071911.2, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064376-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Description
Members of the Ikaros (ZNFN1A1; MIM 603023) family of transcription factors, which includes Pegasus, are expressed in lymphocytes and are implicated in the control of lymphoid development.[supplied by OMIM]
Sequence: GEKKPEPLDFVKDFQEYLTQQTHHVNMISGSVSGDKEAEALQGAGTDGDQNGLDHPSVEVSLDENSGMLVDGFERTFDGKLKCRYCNYASKGTARLIEHSpecifications
NP_071911.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GEKKPEPLDFVKDFQEYLTQQTHHVNMISGSVSGDKEAEALQGAGTDGDQNGLDHPSVEVSLDENSGMLVDGFERTFDGKLKCRYCNYASKGTARLIEH | |
RUO | |
IKZF5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64376 | |
ZNFN1A5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781B0249/FLJ22973/PEGASUS/ZNFN1A5 | |
IKZF5 | |
Recombinant | |
wheat germ expression system |