Learn More
Abnova™ Human ZHX3 Partial ORF (NP_055850.1, 863 a.a. - 955 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00023051-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor. [provided by RefSeq]
Sequence: EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLETSpezifikation
NP_055850.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLET | |
RUO | |
ZHX3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23051 | |
ZHX3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0395/TIX1 | |
ZHX3 | |
Recombinant | |
wheat germ expression system |