Learn More
Abnova™ Human UBE3B Partial ORF (NP_542783, 118 a.a. - 225 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00089910-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E3 ubiquitin-conjugating enzyme family. The encoded protein may interact with other proteins and play a role in stress response. Alternatively spliced transcript variants encoding the same protein isoform have been identified for this gene. [provided by RefSeq]
Sequence: VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLSSpezifikation
NP_542783 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLS | |
RUO | |
UBE3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
89910 | |
UBE3B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586K2123/DKFZp686A1051/FLJ45294/MGC131858/MGC78388 | |
UBE3B | |
Recombinant | |
wheat germ expression system |