Learn More
Abnova™ Human UBE3B Partial ORF (NP_542783, 118 a.a. - 225 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_542783 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 89910 |
Molekulargewicht | 37.62kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16161087
|
Abnova™
H00089910-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16151087
|
Abnova™
H00089910-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E3 ubiquitin-conjugating enzyme family. The encoded protein may interact with other proteins and play a role in stress response. Alternatively spliced transcript variants encoding the same protein isoform have been identified for this gene. [provided by RefSeq]
Sequence: VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLSSpezifikation
NP_542783 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586K2123/DKFZp686A1051/FLJ45294/MGC131858/MGC78388 | |
UBE3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
89910 | |
UBE3B (Human) Recombinant Protein (Q01) | |
VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLS | |
RUO | |
UBE3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |