Learn More
Abnova™ Human TRAPPC3 Full-length ORF (NP_055223.1, 1 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00027095-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
TRAPPC3 is a component of the TRAPP complex, which is involved in tethering of transport vesicles to the cis-Golgi membrane (Turnbull et al., 2005 [PubMed 15692564]).[supplied by OMIM]
Sequence: MSRQANRGTESKKMSSELFTLTYGALVTQLCKDYENDEDVNKQLDKMGFNIGVRLIEDFLARSNVGRCHDFRETADVIAKVAFKMYLGITPSITNWSPAGDEFSLILENNPLVDFVELPDNHSSLIYSNLLCGVLRGALEMVQMAVEAKFVQDTLKGDGVTEIRMRFIRRIEDNLPAGEESpezifikation
NP_055223.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BET3 | |
TRAPPC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27095 | |
TRAPPC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRQANRGTESKKMSSELFTLTYGALVTQLCKDYENDEDVNKQLDKMGFNIGVRLIEDFLARSNVGRCHDFRETADVIAKVAFKMYLGITPSITNWSPAGDEFSLILENNPLVDFVELPDNHSSLIYSNLLCGVLRGALEMVQMAVEAKFVQDTLKGDGVTEIRMRFIRRIEDNLPAGEE | |
RUO | |
TRAPPC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |