missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Tonsil Membrane Tissue Lysate (Adult Membrane Tumor)

Artikelnummer. 18210451 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0,1 mg
Unit Size:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18210451 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18210451 Supplier Novus Biologicals™ Supplier No. NBL126441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Human Tonsil Membrane Tissue Lysate (Adult Membrane Tumor) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Lymph
Menge 0,1 mg
Lagerungspuffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Testspezifität Membrane Protein - Human Tumor Tissue: Tonsil
Zelluläre Fraktion Membrane
Gewebezustand Tumor
Kreuzreaktivität Human
Wirtsspezies Human
Produkttyp Human Tonsil Membrane Tissue Lysate (Adult Tumor)
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Lebensphase Erwachs.
Protein State Native
Show More Show Less

Nur für Forschungszwecke

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.