Learn More
Abnova™ Human TMLHE Full-length ORF (AAH25269.1, 1 a.a. - 376 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00055217-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Epsilon-N-trimethyllysine hydroxylase (EC 1.14.11.8) catalyzes the conversion of epsilon-N-trimethyllysine to beta-hydroxy-N-epsilon-trimethyllysine in the first step of L-carnitine biosynthesis (Vaz et al., 2001 [PubMed 11431483]).[supplied by OMIM]
Sequence: MWYHRLSHLHSRLQDLLKGGVIYPALPQPNFKSLLPLAVHWHHTASKSLTCAWQQHEDHFELKYANTVMRFDYVWLRDHCRSASCYNSKTHQRSLDTASVDLCIKPKTIRLDETTLFFTWPDGHVTKYDLNWLVKNSYEGQKQKVIQPRILWNAEIYQQAQVPSVDCQSFLETNEGLKKFLQNFLLYGIAFVENVPPTQEHTEKLAERISLIRETIYGRMWYFTSDFSRGDTAYTKLALDRHTDTTYFQEPCGIQVFHCLKHEGTGGRTLLVDGFYAAEQVLQKAPEEFELLSKVPLKHEYIEDVGECHNHMIGIGPVLNIYPWNKELYLIRLFKEKQNTVNRQWNSSLQCDIPERILTYRHFVSGTSIEHRGSLISpezifikation
AAH25269.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BBOX2/FLJ10727/TMLH/XAP130 | |
TMLHE | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55217 | |
TMLHE (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWYHRLSHLHSRLQDLLKGGVIYPALPQPNFKSLLPLAVHWHHTASKSLTCAWQQHEDHFELKYANTVMRFDYVWLRDHCRSASCYNSKTHQRSLDTASVDLCIKPKTIRLDETTLFFTWPDGHVTKYDLNWLVKNSYEGQKQKVIQPRILWNAEIYQQAQVPSVDCQSFLETNEGLKKFLQNFLLYGIAFVENVPPTQEHTEKLAERISLIRETIYGRMWYFTSDFSRGDTAYTKLALDRHTDTTYFQEPCGIQVFHCLKHEGTGGRTLLVDGFYAAEQVLQKAPEEFELLSKVPLKHEYIEDVGECHNHMIGIGPVLNIYPWNKELYLIRLFKEKQNTVNRQWNSSLQCDIPERILTYRHFVSGTSIEHRGSLI | |
RUO | |
TMLHE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |