Learn More
Abnova™ Human TMED1 Partial ORF (NP_006849.1, 50 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00011018-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known. [provided by RefSeq]
Sequence: APANASLETEYQVIGGAGLDVDFTLESPQGVLLVSESRKADGVHTVEPTEAGDYKLCFDNSFSTISEKLVFFELIFDSLQDDEEVEGWAEAVEPEEMLDSpezifikation
NP_006849.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
APANASLETEYQVIGGAGLDVDFTLESPQGVLLVSESRKADGVHTVEPTEAGDYKLCFDNSFSTISEKLVFFELIFDSLQDDEEVEGWAEAVEPEEMLD | |
RUO | |
TMED1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11018 | |
TMED1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL1RL1LG/Il1rl1l/MGC1270/ST2L | |
TMED1 | |
Recombinant | |
wheat germ expression system |