Learn More
Abnova™ Human TLX3 Full-length ORF (NP_066305.2, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00030012-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
RNX (HOX11L2, TLX3) belongs to a family of orphan homeobox genes that encode DNA-binding nuclear transcription factors. Members of the HOX11 gene family are characterized by a threonine-47 replacing cytosine in the highly conserved homeodomain (Dear et al., 1993 [PubMed 8099440]).[supplied by OMIM]
Sequence: MEAPASAQTPHPHEPISFGIDQILNSPDQDSAPAPRGPDGASYLGGPPGGRPGATYPSLPASFAGLGAPFEDAGSYSVNLSLAPAGVIRVPAHRPLPGAVPPPLPSALPAMPSVPTVSSLGGLNFPWMESSRRFVKDRFTAAAALTPFTVTRRIGHPYQNRTPPKRKKPRTSFSRVQICELEKRFHRQKYLASAERAALAKSLKMTDAQVKTWFQNRRTKWRRQTAEEREAERQQASRLMLQLQHDAFQKSLNDSIQPDPLCLHNSSLFALQNLQPWEEDSSKVPAVTSLVSpezifikation
NP_066305.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX11L2/MGC29804/RNX | |
TLX3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
30012 | |
TLX3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEAPASAQTPHPHEPISFGIDQILNSPDQDSAPAPRGPDGASYLGGPPGGRPGATYPSLPASFAGLGAPFEDAGSYSVNLSLAPAGVIRVPAHRPLPGAVPPPLPSALPAMPSVPTVSSLGGLNFPWMESSRRFVKDRFTAAAALTPFTVTRRIGHPYQNRTPPKRKKPRTSFSRVQICELEKRFHRQKYLASAERAALAKSLKMTDAQVKTWFQNRRTKWRRQTAEEREAERQQASRLMLQLQHDAFQKSLNDSIQPDPLCLHNSSLFALQNLQPWEEDSSKVPAVTSLV | |
RUO | |
TLX3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |