missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-9 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Marke: R&D Systems FAB1139A-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.
Spezifikation
| Siglec-9 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27180 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 191240 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9Y336 | |
| SIGLEC9 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
| 100 Tests | |
| Primary | |
| Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur