missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Serpin H1 Protein, His Tag
403.52 CHF - 2673.33 CHF
Spezifikation
Zugriffsnummer | NP_001226.2 |
---|---|
Molekulargewicht | 45.5 kDa |
Quelle | Human Serpin H1, His Tag (SE1-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 418 (Accession # P50454-1). |
Immunogen | Ala 19 - Leu 418 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15837667
|
ACROBiosystems
SE1-H5224-50UG |
50 μg |
403.52 CHF
50 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827667
|
ACROBiosystems
SE1-H5224-1MG |
1 mg |
2673.33 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Serpin H1 is also known as SERPINH1, 47 kDa heat shock protein (HSP47), Collagen-binding protein (Colligin), Arsenic-transactivated protein 3 (AsTP3), which belongs to the serpin family. SERPINH1 binds specifically to collagen. SERPINH1 / HSP47 could be involved as a chaperone in the biosynthetic pathway of collagen.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.5 kDa. The protein migrates as 47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001226.2 | |
Human Serpin H1, His Tag (SE1-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 418 (Accession # P50454-1). | |
-20°C | |
Serpin H1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
45.5 kDa | |
Ala 19 - Leu 418 | |
SERPINH1,Serpin H1,HSP47,CBP1,CBP2,SERPINH2,PIG14,AsTP3,Colligin,OI10,PPROM,RA-A47,gp46 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |