Learn More
ACROBiosystems Human Serpin H1 Protein, His Tag
Marke: ACROBiosystems SE1-H5224-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Serpin H1 is also known as SERPINH1, 47 kDa heat shock protein (HSP47), Collagen-binding protein (Colligin), Arsenic-transactivated protein 3 (AsTP3), which belongs to the serpin family. SERPINH1 binds specifically to collagen. SERPINH1 / HSP47 could be involved as a chaperone in the biosynthetic pathway of collagen.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.5 kDa. The protein migrates as 47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001226.2 | |
1 mg | |
Ala 19 - Leu 418 | |
SERPINH1,Serpin H1,HSP47,CBP1,CBP2,SERPINH2,PIG14,AsTP3,Colligin,OI10,PPROM,RA-A47,gp46 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
45.5 kDa | |
Human Serpin H1, His Tag (SE1-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 418 (Accession # P50454-1). | |
-20°C | |
Serpin H1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |