missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC14L4 Full-length ORF (CAK54647.1, 1 a.a. - 352 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00284904-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129983

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 13-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQ
Spezifikation

Spezifikation

CAK54647.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.12kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAP3
SEC14L4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
284904
SEC14L4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLRRHMEFRKQQDLDNIVTWQPPEVIQLYDSGGLCGYDYEGCPVYFNIIGSLDPKGLLLSASKQDMIRKRIKVCELLLHECELQTQKLGRKIEMALMVFDMEGLSLKHLWKPAVEVYQQFFSILEANYPETLKNLIVIRAPKLFPVAFNLVKSFMSEETRRKIVILGDNWKQELTKFISPDQLPVEFGGTMTDPDGNPKCLTKINYGGEVPKSYYLCEQVRLQYEHTRSVGRGSSLQVENEILFPGCVLRWQFASDGGDIGFGVFLKTKMGEQQSAREMTEVLPSQRYNAHMVPEDGSLTCLQAGVYVLRFDNTYSRMHAKKLSYTVEVLLPDKASEETLQSLKAMRPSPTQ
RUO
SEC14L4
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote