missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Prokineticin-2 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marke: Gibco™ 100-46-20UG
Additional Details : Gewicht : 0.16500kg
Beschreibung
Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.Spezifikation
Q9HC23 | |
protein with no preservative | |
8.8 | |
20 μg | |
-20°C | |
<1 EU/μg | |
PROK2 | |
Unconjugated | |
AVITGACDKD SQCGGGMCCA VSIWVKSIRI CTPMGKLGDS CHPLTRKVPF FGRRMHHTCP CLPGLACLRT SFNRFICLAQ K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
60675 | |
Human Prokineticin-2 | |
E. coli | |
RUO | |
Bombina variegata 8 kD protein; Bombina variegata 8 kDa protein; Bv 8 homolog (Bombina variegata); BV8; Bv8/prokineticin 2-like protein; Bv8/prokineticin 2-like protein splice variant; HH4; homolog of mouse Bv8 (Bombina variegata 8 kDa); homolog of mouse Bv8 (Bombina variegata 8 kDa); prokineticin 2; KAL4; MIT1; PK2; Prok1; Prok2; prokineticin 1; prokineticin 2; prokineticin 2 beta; prokineticin-2; Protein Bv8 homolog; secreted protein | |
Human | |
Recombinant | |
Lyophilized |