Learn More
Abnova™ Human PRKAG3 Full-length ORF (NP_059127.2, 1 a.a. - 489 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00053632-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. It is dominantly expressed in skeletal muscle. Studies of the pig counterpart suggest that this subunit may play a key role in the regulation of energy metabolism in skeletal muscle. [provided by RefSeq]
Sequence: MEPGLEHALRRTPSWSSLGGSEHQEMSFLEQENSSSWPSPAVTSSSERIRGKRRAKALRWTRQKSVEEGEPPGQGEGPRSRPAAESTGLEATFPKTTPLAQADPAGVGTPPTGWDCLPSDCTASAAGSSTDDVELATEFPATEAWECELEGLLEERPALCLSPQAPFPKLGWDDELRKPGAQIYMRFMQEHTCYDAMATSSKLVIFDTMLEIKKAFFALVANGVRAAPLWDSKKQSFVGMLTITDFILVLHRYYRSPLVQIYEIEQHKIETWREIYLQGCFKPLVSISPNDSLFEAVYTLIKNRIHRLPVLDPVSGNVLHILTHKRLLKFLHIFGSLLPRPSFLYRTIQDLGIGTFRDLAVVLETAPILTALDIFVDRRVSALPVVNECGQVVGLYSRFDVIHLAAQQTYNHLDMSVGEALRQRTLCLEGVLSCQPHESLGEVIDRIAREQVHRLVLVDETQHLLGVVSLSDILQALVLSPAGIDALGASpezifikation
NP_059127.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMPKG3 | |
PRKAG3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53632 | |
PRKAG3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPGLEHALRRTPSWSSLGGSEHQEMSFLEQENSSSWPSPAVTSSSERIRGKRRAKALRWTRQKSVEEGEPPGQGEGPRSRPAAESTGLEATFPKTTPLAQADPAGVGTPPTGWDCLPSDCTASAAGSSTDDVELATEFPATEAWECELEGLLEERPALCLSPQAPFPKLGWDDELRKPGAQIYMRFMQEHTCYDAMATSSKLVIFDTMLEIKKAFFALVANGVRAAPLWDSKKQSFVGMLTITDFILVLHRYYRSPLVQIYEIEQHKIETWREIYLQGCFKPLVSISPNDSLFEAVYTLIKNRIHRLPVLDPVSGNVLHILTHKRLLKFLHIFGSLLPRPSFLYRTIQDLGIGTFRDLAVVLETAPILTALDIFVDRRVSALPVVNECGQVVGLYSRFDVIHLAAQQTYNHLDMSVGEALRQRTLCLEGVLSCQPHESLGEVIDRIAREQVHRLVLVDETQHLLGVVSLSDILQALVLSPAGIDALGA | |
RUO | |
PRKAG3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |