Learn More
Abnova™ Human PPFIA3 Partial ORF (NP_003651, 1043 a.a. - 1139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008541-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family. Liprins interact with members of LAR family of transmembrane protein tyrosine phosphatases, which are known to be important for axon guidance and mammary gland development. Liprin family protein has been shown to localize phosphatase LAR to cell focal adhesions and may be involved in the molecular organization of presynaptic active zones. [provided by RefSeq]
Sequence: NERVMGWVSGLGLKEFATNLTESGVHGALLALDETFDYSDLALLLQIPTQNAQARQLLEKEFSNLISLGTDRRLDEDSAKSFSRSPSWRKMFREKDLSpezifikation
NP_003651 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NERVMGWVSGLGLKEFATNLTESGVHGALLALDETFDYSDLALLLQIPTQNAQARQLLEKEFSNLISLGTDRRLDEDSAKSFSRSPSWRKMFREKDL | |
RUO | |
PPFIA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8541 | |
PPFIA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0654/LPNA3/MGC126567/MGC126569 | |
PPFIA3 | |
Recombinant | |
wheat germ expression system |