Learn More
Abnova™ Human POMT2 Full-length ORF (NP_037514.2, 1 a.a. - 750 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00029954-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS)
Sequence: MPPATGGGLAESELRPRRGRCGPQAARAAGRDVAAEAVARSPKRPAWGSRRFEAVGWWALLALVTLLSFATRFHRLDEPPHICWDETHFGKMGSYYINRTFFFDVHPPLGKMLIGLAGYLSGYDGTFLFQKPGDKYEHHSYMGMRGFCAFLGSWLVPFAYLTVLDLSKSLSAALLTAALLTFDTGCLTLSQYILLDPILMFFIMAAMLSMVKYNSCADRPFSAPWWFWLSLTGVSLAGALGVKFVGLFIILQVGLNTIADLWYLFGDLSLSLVTVGKHLTARVLCLIVLPLALYTATFAVHFMVLSKSGPGDGFFSSAFQARLSGNNLHNASIPEHLAYGSVITVKNLRMAIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPSFPVEFVRHGDIIRLEHKETSRNLHSHYHEAPMTRKHYQVTGYGINGTGDSNDFWRIEVVNRKFGNRIKVLRSRIRFIHLVTGCVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFSGVNDTDFRVYLLGNPVVWWLNLLSIALYLLSGSIIAVAMQRGARLPAEVAGLSQVLLRGGGQVLLGWTLHYFPFFLMGRVLYFHHYFPAMLFSSMLTGILWDTLLRLCAWGLASWPLARGIHVAGILSLLLGTAYSFYLFHPLAYGMVGPLAQDPQSPMAGLRWLDSWDFSpezifikation
NP_037514.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G10254/FLJ22309 | |
POMT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
29954 | |
POMT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPPATGGGLAESELRPRRGRCGPQAARAAGRDVAAEAVARSPKRPAWGSRRFEAVGWWALLALVTLLSFATRFHRLDEPPHICWDETHFGKMGSYYINRTFFFDVHPPLGKMLIGLAGYLSGYDGTFLFQKPGDKYEHHSYMGMRGFCAFLGSWLVPFAYLTVLDLSKSLSAALLTAALLTFDTGCLTLSQYILLDPILMFFIMAAMLSMVKYNSCADRPFSAPWWFWLSLTGVSLAGALGVKFVGLFIILQVGLNTIADLWYLFGDLSLSLVTVGKHLTARVLCLIVLPLALYTATFAVHFMVLSKSGPGDGFFSSAFQARLSGNNLHNASIPEHLAYGSVITVKNLRMAIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPSFPVEFVRHGDIIRLEHKETSRNLHSHYHEAPMTRKHYQVTGYGINGTGDSNDFWRIEVVNRKFGNRIKVLRSRIRFIHLVTGCVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFSGVNDTDFRVYLLGNPVVWWLNLLSIALYLLSGSIIAVAMQRGARLPAEVAGLSQVLLRGGGQVLLGWTLHYFPFFLMGRVLYFHHYFPAMLFSSMLTGILWDTLLRLCAWGLASWPLARGIHVAGILSLLLGTAYSFYLFHPLAYGMVGPLAQDPQSPMAGLRWLDSWDF | |
RUO | |
POMT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |