missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human POMT2 Full-length ORF (NP_037514.2, 1 a.a. - 750 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_037514.2 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 29954 |
Molekulargewicht | 110.6kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16129242
|
Abnova™
H00029954-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16119242
|
Abnova™
H00029954-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS)
Sequence: MPPATGGGLAESELRPRRGRCGPQAARAAGRDVAAEAVARSPKRPAWGSRRFEAVGWWALLALVTLLSFATRFHRLDEPPHICWDETHFGKMGSYYINRTFFFDVHPPLGKMLIGLAGYLSGYDGTFLFQKPGDKYEHHSYMGMRGFCAFLGSWLVPFAYLTVLDLSKSLSAALLTAALLTFDTGCLTLSQYILLDPILMFFIMAAMLSMVKYNSCADRPFSAPWWFWLSLTGVSLAGALGVKFVGLFIILQVGLNTIADLWYLFGDLSLSLVTVGKHLTARVLCLIVLPLALYTATFAVHFMVLSKSGPGDGFFSSAFQARLSGNNLHNASIPEHLAYGSVITVKNLRMAIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPSFPVEFVRHGDIIRLEHKETSRNLHSHYHEAPMTRKHYQVTGYGINGTGDSNDFWRIEVVNRKFGNRIKVLRSRIRFIHLVTGCVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFSGVNDTDFRVYLLGNPVVWWLNLLSIALYLLSGSIIAVAMQRGARLPAEVAGLSQVLLRGGGQVLLGWTLHYFPFFLMGRVLYFHHYFPAMLFSSMLTGILWDTLLRLCAWGLASWPLARGIHVAGILSLLLGTAYSFYLFHPLAYGMVGPLAQDPQSPMAGLRWLDSWDFSpezifikation
NP_037514.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPPATGGGLAESELRPRRGRCGPQAARAAGRDVAAEAVARSPKRPAWGSRRFEAVGWWALLALVTLLSFATRFHRLDEPPHICWDETHFGKMGSYYINRTFFFDVHPPLGKMLIGLAGYLSGYDGTFLFQKPGDKYEHHSYMGMRGFCAFLGSWLVPFAYLTVLDLSKSLSAALLTAALLTFDTGCLTLSQYILLDPILMFFIMAAMLSMVKYNSCADRPFSAPWWFWLSLTGVSLAGALGVKFVGLFIILQVGLNTIADLWYLFGDLSLSLVTVGKHLTARVLCLIVLPLALYTATFAVHFMVLSKSGPGDGFFSSAFQARLSGNNLHNASIPEHLAYGSVITVKNLRMAIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPSFPVEFVRHGDIIRLEHKETSRNLHSHYHEAPMTRKHYQVTGYGINGTGDSNDFWRIEVVNRKFGNRIKVLRSRIRFIHLVTGCVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFSGVNDTDFRVYLLGNPVVWWLNLLSIALYLLSGSIIAVAMQRGARLPAEVAGLSQVLLRGGGQVLLGWTLHYFPFFLMGRVLYFHHYFPAMLFSSMLTGILWDTLLRLCAWGLASWPLARGIHVAGILSLLLGTAYSFYLFHPLAYGMVGPLAQDPQSPMAGLRWLDSWDF | |
RUO | |
POMT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
29954 | |
POMT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G10254/FLJ22309 | |
POMT2 | |
Recombinant | |
wheat germ expression system |