Learn More
Abnova™ Human POLR1B Partial ORF (NP_061887, 963 a.a. - 1072 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084172-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Eukaryotic RNA polymerase I (pol I) is responsible for the transcription of ribosomal RNA (rRNA) genes and production of rRNA, the primary component of ribosomes. Pol I is a multisubunit enzyme composed of 6 to 14 polypeptides, depending on the species. Most of the mass of the pol I complex derives from the 2 largest subunits, Rpa1 and Rpa2 in yeast. POLR1B is homologous to Rpa2 (Seither and Grummt, 1996 [PubMed 8921381]).[supplied by OMIM]
Sequence: GEMLKAAGYNFYGTERLYSGISGLELEADIFIGVVYYQRLRHMVSDKFQVRTTGARDRVTNQPIGGRNVQGGIRFGEMERDALLAHGTSFLLHDRLFNCSDRSVAHVCVKSpezifikation
NP_061887 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GEMLKAAGYNFYGTERLYSGISGLELEADIFIGVVYYQRLRHMVSDKFQVRTTGARDRVTNQPIGGRNVQGGIRFGEMERDALLAHGTSFLLHDRLFNCSDRSVAHVCVK | |
RUO | |
POLR1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84172 | |
POLR1B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10816/FLJ21921/MGC131780/RPA135/RPA2/Rpo1-2 | |
POLR1B | |
Recombinant | |
wheat germ expression system |