Learn More
Abnova™ Human PHF10 Full-length ORF (NP_060758.1, 1 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00055274-P02.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28. [provided by RefSeq]
Sequence: MLQEQVSEYLGVTSFKRKYPDLERRDLSHKEKLYLRELNVITETQCTLGLTALRSDEVIDLMIKEYPAKHAEYSVILQEKERQRITDHYKEYSQMQQQNTQKVEASKVPEYIKKAAKKAAEFNSNLNRERMEERRAYFDLQTHVIQVPQGKYKVLPTERTKVSSYPVALIPGQFQEYYKRYSPDELRYLPLNTALYEPPLDPELPALDSDGDSDDGEDGRGDEKRKNKGTSDSSSGNVSEGESPPDSQEDSFQGRQKSKDKAATPRKDGPKRSVLSKSVPGYKPKVIPNAICGICLKGKESNKKGKAESLIHCSQCENSGHPSCLDMTMELVSMIKTYPWQCMECKTCIICGQPHHEEEMMFCDMCDRGYHTFCVGLGAIPSGRWICDCCQRAPPTPRKVGRRGKNSKEGSpezifikation
NP_060758.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10975/MGC111009/XAP135 | |
PHF10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55274 | |
PHF10 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLQEQVSEYLGVTSFKRKYPDLERRDLSHKEKLYLRELNVITETQCTLGLTALRSDEVIDLMIKEYPAKHAEYSVILQEKERQRITDHYKEYSQMQQQNTQKVEASKVPEYIKKAAKKAAEFNSNLNRERMEERRAYFDLQTHVIQVPQGKYKVLPTERTKVSSYPVALIPGQFQEYYKRYSPDELRYLPLNTALYEPPLDPELPALDSDGDSDDGEDGRGDEKRKNKGTSDSSSGNVSEGESPPDSQEDSFQGRQKSKDKAATPRKDGPKRSVLSKSVPGYKPKVIPNAICGICLKGKESNKKGKAESLIHCSQCENSGHPSCLDMTMELVSMIKTYPWQCMECKTCIICGQPHHEEEMMFCDMCDRGYHTFCVGLGAIPSGRWICDCCQRAPPTPRKVGRRGKNSKEG | |
RUO | |
PHF10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |