Learn More
Abnova™ Human PCDH10 Partial ORF (NP_065866, 18 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00057575-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains. [provided by RefSeq]
Sequence: SQLHYTVQEEQEHGTFVGNIAEDLGLDITKLSARGFQTVPNSRTPYLDLNLETGVLYVNEKIDREQICKQSPSCVLHLEVFLENPLELFQVEIEVLDINDNPPSFPEPDLSpezifikation
NP_065866 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SQLHYTVQEEQEHGTFVGNIAEDLGLDITKLSARGFQTVPNSRTPYLDLNLETGVLYVNEKIDREQICKQSPSCVLHLEVFLENPLELFQVEIEVLDINDNPPSFPEPDL | |
RUO | |
PCDH10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57575 | |
PCDH10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761O2023/KIAA1400/MGC133344/OL-PCDH/PCDH19 | |
PCDH10 | |
Recombinant | |
wheat germ expression system |