missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDH10 Partial ORF (NP_065866, 18 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00057575-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129186

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains. [provided by RefSeq]

Sequence: SQLHYTVQEEQEHGTFVGNIAEDLGLDITKLSARGFQTVPNSRTPYLDLNLETGVLYVNEKIDREQICKQSPSCVLHLEVFLENPLELFQVEIEVLDINDNPPSFPEPDL
Spezifikation

Spezifikation

NP_065866
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SQLHYTVQEEQEHGTFVGNIAEDLGLDITKLSARGFQTVPNSRTPYLDLNLETGVLYVNEKIDREQICKQSPSCVLHLEVFLENPLELFQVEIEVLDINDNPPSFPEPDL
RUO
PCDH10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57575
PCDH10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp761O2023/KIAA1400/MGC133344/OL-PCDH/PCDH19
PCDH10
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote