Learn More
Abnova™ Human PAPPA Partial ORF (NP_002572, 85 a.a. - 194 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005069-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses. [provided by RefSeq]
Sequence: GATEEPSPPSRALYFSGRGEQLRLRADLELPRDAFTLQVWLRAEGGQRSPAVITGLYDKCSYISRDRGWVVGIHTISDQDNKDPRYFFSLKTDRARQVTTINAHRSYLPGSpezifikation
NP_002572 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GATEEPSPPSRALYFSGRGEQLRLRADLELPRDAFTLQVWLRAEGGQRSPAVITGLYDKCSYISRDRGWVVGIHTISDQDNKDPRYFFSLKTDRARQVTTINAHRSYLPG | |
RUO | |
PAPPA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5069 | |
PAPPA (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASBABP2/DIPLA1/IGFBP-4ase/PAPA/PAPP-A/PAPPA1 | |
PAPPA | |
Recombinant | |
wheat germ expression system |