Learn More
Abnova™ Human PAFAH2 Full-length ORF (NP_000428.2, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005051-P01.25ug
Additional Details : Gewicht : 0.00010kg
Description
This gene encodes platelet-activating factor acetylhydrolase isoform 2, a single-subunit intracellular enzyme that catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). However, this lipase exhibits a broader substrate specificity than simply platelet activating factor. Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist, and both are multi-subunit enzymes. Additionally, there is a single-subunit serum isoform of this enzyme. [provided by RefSeq]
Sequence: MGVNQSVGFPPVTGPHLVGCGDVMEGQNLQGSFFRLFYPCQKAEETMEQPLWIPRYEYCTGLAEYLQFNKRCGGLLFNLAVGSCRLPVSWNGPFKTKDSGYPLIIFSHGLGAFRTLYSAFCMELASRGFVVAVPEHRDRSAATTYFCKQAPEENQPTNESLQEEWIPFRRVEEGEKEFHVRNPQVHQRVSECLRVLKILQEVTAGQTVFNILPGGLDLMTLKGNIDMSRVAVMGHSFGGATAILALAKETQFRCAVALDAWMFPLERDFYPKARGPVFFINTEKFQTMESVNLMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSLSpecifications
NP_000428.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26025/HSD-PLA2 | |
PAFAH2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5051 | |
PAFAH2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGVNQSVGFPPVTGPHLVGCGDVMEGQNLQGSFFRLFYPCQKAEETMEQPLWIPRYEYCTGLAEYLQFNKRCGGLLFNLAVGSCRLPVSWNGPFKTKDSGYPLIIFSHGLGAFRTLYSAFCMELASRGFVVAVPEHRDRSAATTYFCKQAPEENQPTNESLQEEWIPFRRVEEGEKEFHVRNPQVHQRVSECLRVLKILQEVTAGQTVFNILPGGLDLMTLKGNIDMSRVAVMGHSFGGATAILALAKETQFRCAVALDAWMFPLERDFYPKARGPVFFINTEKFQTMESVNLMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSL | |
RUO | |
PAFAH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |