missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PAFAH2 Full-length ORF (NP_000428.2, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00005051-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16151028

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 14-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes platelet-activating factor acetylhydrolase isoform 2, a single-subunit intracellular enzyme that catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). However, this lipase exhibits a broader substrate specificity than simply platelet activating factor. Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist, and both are multi-subunit enzymes. Additionally, there is a single-subunit serum isoform of this enzyme. [provided by RefSeq]

Sequence: MGVNQSVGFPPVTGPHLVGCGDVMEGQNLQGSFFRLFYPCQKAEETMEQPLWIPRYEYCTGLAEYLQFNKRCGGLLFNLAVGSCRLPVSWNGPFKTKDSGYPLIIFSHGLGAFRTLYSAFCMELASRGFVVAVPEHRDRSAATTYFCKQAPEENQPTNESLQEEWIPFRRVEEGEKEFHVRNPQVHQRVSECLRVLKILQEVTAGQTVFNILPGGLDLMTLKGNIDMSRVAVMGHSFGGATAILALAKETQFRCAVALDAWMFPLERDFYPKARGPVFFINTEKFQTMESVNLMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSL
Spezifikation

Spezifikation

NP_000428.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ26025/HSD-PLA2
PAFAH2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5051
PAFAH2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGVNQSVGFPPVTGPHLVGCGDVMEGQNLQGSFFRLFYPCQKAEETMEQPLWIPRYEYCTGLAEYLQFNKRCGGLLFNLAVGSCRLPVSWNGPFKTKDSGYPLIIFSHGLGAFRTLYSAFCMELASRGFVVAVPEHRDRSAATTYFCKQAPEENQPTNESLQEEWIPFRRVEEGEKEFHVRNPQVHQRVSECLRVLKILQEVTAGQTVFNILPGGLDLMTLKGNIDMSRVAVMGHSFGGATAILALAKETQFRCAVALDAWMFPLERDFYPKARGPVFFINTEKFQTMESVNLMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSL
RUO
PAFAH2
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote