missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human P11 Partial ORF (NP_006016.1, 270 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008909-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16119635

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a protein with protease activity. Its specific function has not been determined. The protein may be useful as a tumor marker. [provided by RefSeq]

Sequence: EGLVDYYSHIYDGPWDSYPDVLAMQFNWDGYYKEVGSAFIGSSPEFEFALYSLCFIARPGKVCQLSLGGYPLAVRTYTWDKSTYGNGKKYIATAYIVSST
Spezifikation

Spezifikation

NP_006016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGLVDYYSHIYDGPWDSYPDVLAMQFNWDGYYKEVGSAFIGSSPEFEFALYSLCFIARPGKVCQLSLGGYPLAVRTYTWDKSTYGNGKKYIATAYIVSST
RUO
P11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8909
P11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC133268/PP11/PRSS26
P11
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote