Learn More
Abnova™ Human OVOL1 Full-length ORF (NP_004552.2, 1 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005017-P02.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined. [provided by RefSeq]
Sequence: MPRAFLVKKPCVSTCKRNWSELPDEERGEIYVPVSLGFCPPQPYREPEPSVAEPPSCPLALNMSLRDSSYSMAPGPCVVAQLPSEDMGHLTDPQSRDHGFLRTKMKVTLGDSPSGDLFTCRVCQKAFTYQRMLNRHMKCHNDVKRHLCTYCGKGFNDTFDLKRHVRTHTGVRPYKCSLCDKAFTQRCSLESHLKKIHGVQQKYAYKERRAKLYVCEECGCTSESQEGHVLHLKEHHPDSPLLRKTSKKVAVALQNTVTSLLQGSPHLSpezifikation
NP_004552.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOVO1 | |
OVOL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5017 | |
OVOL1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRAFLVKKPCVSTCKRNWSELPDEERGEIYVPVSLGFCPPQPYREPEPSVAEPPSCPLALNMSLRDSSYSMAPGPCVVAQLPSEDMGHLTDPQSRDHGFLRTKMKVTLGDSPSGDLFTCRVCQKAFTYQRMLNRHMKCHNDVKRHLCTYCGKGFNDTFDLKRHVRTHTGVRPYKCSLCDKAFTQRCSLESHLKKIHGVQQKYAYKERRAKLYVCEECGCTSESQEGHVLHLKEHHPDSPLLRKTSKKVAVALQNTVTSLLQGSPHL | |
RUO | |
OVOL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |