missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ODF2L Full-length ORF (AAH91490.1, 1 a.a. - 620 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00057489-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MEKAVNDGSHSEELFCHLKTISEKEDLPRCTSESHLSCLKQDILNEKTELEATLKEAELVTHSVELLLPLFKDTIEKINFENANLSALNLKISEQKEILIKELDTFKSVKLALEHLLRKRDYKQTGDNLSSMLLENLTDNESENTNLKKKVFEKEAHIQELSCLFQSEKANTLKANRFSQSVKVVHERLQIQIHKREAENDKLKEYVKSLETKIAKWNLQSRMNKNEAIVMKEASRQKTVALKKASKVYKQRLDHFTGAIEKLTSQIRDQEAKLSETISASNAWKSHYEKIVIEKTELEVQIETMKKQIINLLEDLKKMEDHGKNSCEEILRKVHSIEYENETLNLENTKLKLRFPCRITESKNMNILIVLDMLCYISSEKTTLAALKDEVVSVENELSELQEVEKKQKTLIEMYKTQVQKLQEAAEIVKSRCENLLHKNNQITKTKNKNVEKVDGNHNLLTKLSLEEENCLIQLKCENLQQKLEQMDAENKELEKKLANQEECLKHSNLKFKEKSAEYTALARQLEAALEEGRQKVAEEIEKMSSRESALQIKILDLETELRKKNEEQNQLVCKMNSKAQHQEVCLKEVQNSLEKSENQNESIKNYLQFLKTSYVTMFESpezifikation
AAH91490.1 | |
Liquid | |
57489 | |
ODF2L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKAVNDGSHSEELFCHLKTISEKEDLPRCTSESHLSCLKQDILNEKTELEATLKEAELVTHSVELLLPLFKDTIEKINFENANLSALNLKISEQKEILIKELDTFKSVKLALEHLLRKRDYKQTGDNLSSMLLENLTDNESENTNLKKKVFEKEAHIQELSCLFQSEKANTLKANRFSQSVKVVHERLQIQIHKREAENDKLKEYVKSLETKIAKWNLQSRMNKNEAIVMKEASRQKTVALKKASKVYKQRLDHFTGAIEKLTSQIRDQEAKLSETISASNAWKSHYEKIVIEKTELEVQIETMKKQIINLLEDLKKMEDHGKNSCEEILRKVHSIEYENETLNLENTKLKLRFPCRITESKNMNILIVLDMLCYISSEKTTLAALKDEVVSVENELSELQEVEKKQKTLIEMYKTQVQKLQEAAEIVKSRCENLLHKNNQITKTKNKNVEKVDGNHNLLTKLSLEEENCLIQLKCENLQQKLEQMDAENKELEKKLANQEECLKHSNLKFKEKSAEYTALARQLEAALEEGRQKVAEEIEKMSSRESALQIKILDLETELRKKNEEQNQLVCKMNSKAQHQEVCLKEVQNSLEKSENQNESIKNYLQFLKTSYVTMFE | |
RUO | |
ODF2L | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
98.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1229/MGC111060/RP5-977L11.1/dJ977L11.1 | |
ODF2L | |
Yes | |
wheat germ expression system |