missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYO7A Partial ORF (NP_000251, 2118 a.a. - 2213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00004647-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16121615

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 14-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene is a member of the myosin gene family. Myosins are mechanochemical proteins characterized by the presence of a motor domain, an actin-binding domain, a neck domain that interacts with other proteins, and a tail domain that serves as an anchor. This gene encodes an unconventional myosin with a very short tail. Defects in this gene are associated with the mouse shaker-1 phenotype and the human Usher syndrome 1B which are characterized by deafness, reduced vestibular function, and (in human) retinal degeneration. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: KQTTEPNFPEILLIAINKYGVSLIDPKTKDILTTHPFTKISNWSSGNTYFHITIGNLVRGSKLLCETSLGYKMDDLLTSYISQMLTAMSKQRGSRS
Spezifikation

Spezifikation

NP_000251
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KQTTEPNFPEILLIAINKYGVSLIDPKTKDILTTHPFTKISNWSSGNTYFHITIGNLVRGSKLLCETSLGYKMDDLLTSYISQMLTAMSKQRGSRS
RUO
MYO7A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4647
MYO7A (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNA11/DFNB2/MYOVIIA/MYU7A/NSRD2/USH1B
MYO7A
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote