missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse VGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
245.00 CHF - 529.00 CHF
Spezifikation
| Antigen | VGF |
|---|---|
| Klon | 619707 |
| Konzentration | LYOPH |
| Verdünnung | Immunohistochemistry 8-25 ug/mL |
| Anwendungen | Immunohistochemistry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18746929
![]() |
R&D Systems
MAB6317-SP |
25 μg | |||||||
|
18111671
|
R&D Systems
MAB6317 |
100 μg |
529.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
VGF Monoclonal specifically detects VGF in Human, Mouse samples. It is validated for Immunohistochemistry.Spezifikation
| VGF | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O15240 | |
| 7425 | |
| E. coli-derived recombinant human VGF Thr554-Pro615 Accession # O15240 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human and mouse VGF in direct ELISAs. |
| 619707 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible | |
| VGF | |
| IgG3 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts