Learn More
Human/Mouse VGF Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB6317
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
VGF Monoclonal specifically detects VGF in Human, Mouse samples. It is validated for Immunohistochemistry.
Spezifikation
| VGF | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O15240 | |
| VGF | |
| E. coli-derived recombinant human VGF Thr554-Pro615 Accession # O15240 | |
| 100 μg | |
| Primary | |
| Detects human and mouse VGF in direct ELISAs. | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| 619707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7425 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |