Learn More
Abnova™ Human MKLN1 Partial ORF (NP_037387, 633 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004289-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I (MIM 188060) (Adams et al., 1998 [PubMed 9724633]).[supplied by OMIM]
Sequence: RHCKYLIRKHRFEEKAQVDPLSALKYLQNDLYITVDHSDPEETKEFQLLASALFKSGSDFTALGFSDVDHTYAQRTQLFDTLVNFFPDSMTPPKGNLVDLITLSpezifikation
NP_037387 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RHCKYLIRKHRFEEKAQVDPLSALKYLQNDLYITVDHSDPEETKEFQLLASALFKSGSDFTALGFSDVDHTYAQRTQLFDTLVNFFPDSMTPPKGNLVDLITL | |
RUO | |
MKLN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4289 | |
MKLN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11162/TWA2 | |
MKLN1 | |
Recombinant | |
wheat germ expression system |