missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAPK4 Partial ORF (NP_002738, 42 a.a. - 151 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00005596-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129611

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets. [provided by RefSeq]

Sequence: ACRKVAVKKIALSDARSMKHALREIKIIRRLDHDNIVKVYEVLGPKGTDLQGELFKFSVAYIVQEYMETDLARLLEQGTLAEEHAKLFMYQLLRGLKYIHSANVLHRDLK
Spezifikation

Spezifikation

NP_002738
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ACRKVAVKKIALSDARSMKHALREIKIIRRLDHDNIVKVYEVLGPKGTDLQGELFKFSVAYIVQEYMETDLARLLEQGTLAEEHAKLFMYQLLRGLKYIHSANVLHRDLK
RUO
MAPK4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5596
MAPK4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERK3/Erk4/PRKM4/p63MAPK
MAPK4
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote