missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAPK4 Partial ORF (NP_002738, 42 a.a. - 151 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_002738 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 5596 |
Molekulargewicht | 37.84kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16129611
|
Abnova™
H00005596-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16119611
|
Abnova™
H00005596-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets. [provided by RefSeq]
Sequence: ACRKVAVKKIALSDARSMKHALREIKIIRRLDHDNIVKVYEVLGPKGTDLQGELFKFSVAYIVQEYMETDLARLLEQGTLAEEHAKLFMYQLLRGLKYIHSANVLHRDLKSpezifikation
NP_002738 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERK3/Erk4/PRKM4/p63MAPK | |
MAPK4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5596 | |
MAPK4 (Human) Recombinant Protein (Q01) | |
ACRKVAVKKIALSDARSMKHALREIKIIRRLDHDNIVKVYEVLGPKGTDLQGELFKFSVAYIVQEYMETDLARLLEQGTLAEEHAKLFMYQLLRGLKYIHSANVLHRDLK | |
RUO | |
MAPK4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |