Learn More
Abnova™ Human LUC7L Full-length ORF (NP_060502.1, 1 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00055692-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The LUC7L gene may represent a mammalian heterochromatic gene, encoding a putative RNA-binding protein similar to the yeast Luc7p subunit of the U1 snRNP splicing complex that is normally required for 5-prime splice site selection (Tufarelli et al., 2001 [PubMed 11170747]).[supplied by OMIM]
Sequence: MSAQAQMRALLDQLMGTARDGDETRQRVKFTDDRVCKSHLLDCCPHDILAGTRMDLGECTKIHDLALRADYEIASKERDLFFELDAMDHLESFIAECDRRTELAKKRLAETQEEISAEVSAKAEKVHELNEEIGKLLAKAEQLGAEGNVDESQKILMEVEKVRAKKKEAEEEYRNSMPASSFQQQKLRVCEVCSAYLGLHDNDRRLADHFGGKLHLGFIQIREKLDQLRKTVAEKQEKRNQDRLRRREEREREERLSRRSGSRTRDRRRSRSRDRRRRRSRSTSRERRKLSRSRSRDRHRRHRSRSRSHSRGHRRASRDRSAKYKSpezifikation
NP_060502.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10231/LUC7-LIKE/LUC7B1/SR+89 | |
LUC7L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55692 | |
LUC7L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAQAQMRALLDQLMGTARDGDETRQRVKFTDDRVCKSHLLDCCPHDILAGTRMDLGECTKIHDLALRADYEIASKERDLFFELDAMDHLESFIAECDRRTELAKKRLAETQEEISAEVSAKAEKVHELNEEIGKLLAKAEQLGAEGNVDESQKILMEVEKVRAKKKEAEEEYRNSMPASSFQQQKLRVCEVCSAYLGLHDNDRRLADHFGGKLHLGFIQIREKLDQLRKTVAEKQEKRNQDRLRRREEREREERLSRRSGSRTRDRRRSRSRDRRRRRSRSTSRERRKLSRSRSRDRHRRHRSRSRSHSRGHRRASRDRSAKYK | |
RUO | |
LUC7L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |