missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC285382 Full-length ORF (NP_001020437.1, 1 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00285382-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTVSpezifikation
NP_001020437.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3orf70 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
285382 | |
LOC285382 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTV | |
RUO | |
C3orf70 | |
Recombinant | |
wheat germ expression system |