missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC285382 Full-length ORF (NP_001020437.1, 1 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_001020437.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 285382 |
Molekulargewicht | 54.2kDa |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
16130014
|
Abnova™
H00285382-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16120014
|
Abnova™
H00285382-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
Sequence: MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTVSpecifications
NP_001020437.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSAAASPASERGWKSEKLDEAQALARSCAARRPDFQPCDGLSICATHSHGKCFKLHWCCHLGWCHCKYMYQPMTPVEQLPSTEIPARPREPTNTIQISVSLTEHFLKFASVFQPPLPPDSPRYCMISDLFIDNYQVKCINGKMCYVQKQPAPHSHRMSPEEVSAHDALISKESNTPKIDHCSSPSSSEDSGINAIGAHYVESCDEDTEEGAELSSEEDYSPESSWEPDECTLLSPSQSDLEVIETIETTV | |
RUO | |
C3orf70 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
285382 | |
LOC285382 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3orf70 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |