Learn More
Abnova™ Human LHX2 Full-length ORF (NP_004780.3, 1 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009355-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq]
Sequence: MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLFSpezifikation
NP_004780.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LH2/MGC138390/hLhx2 | |
LHX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9355 | |
LHX2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLF | |
RUO | |
LHX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |