missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LCN12 Full-length ORF (AAH41168.1, 1 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00286256-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16140064

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Members of the lipocalin family, such as LCN12, have a common structure consisting of an 8-stranded antiparallel beta-barrel that forms a cup-shaped ligand-binding pocket or calyx. Lipocalins generally bind small hydrophobic ligands and transport them to specific cells (Suzuki et al., 2004 [PubMed 15363845]).[supplied by OMIM]

Sequence: MRLLCGLWLWLSLLKVLQAQTPTPLPLPPPMQSFQGNQFQGEWFVLGLAGNSFRPEHRALLNAFTATFELSDDGRFEVWNAMTRGQHCDTWSYVLIPAAQPGQFTVDHGVEPGADREETRVVDSDYTQFALMLSRRHTSRLAVLRISLLGRSWLLPPGTLDQFICLGRAQGLSDDNIVFPDVTGNMVHLQACWAVGTGPAGMSLVDPRGAGPSVYPGSSAPACAQGSPGSWVPVLNPGSEPPPAAPGPLSWATSSHPGSPVPGHLLPPQVPCPGPPPPAPPAPGPLSRPTSSHPGSPVLGYLLPPQVPCPGPSPPSGSPVLGHLLPSPIPAHKELGLIPGGALDLSSLPWVAAPA
Spezifikation

Spezifikation

AAH41168.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34753/MGC48935
LCN12
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
286256
LCN12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLLCGLWLWLSLLKVLQAQTPTPLPLPPPMQSFQGNQFQGEWFVLGLAGNSFRPEHRALLNAFTATFELSDDGRFEVWNAMTRGQHCDTWSYVLIPAAQPGQFTVDHGVEPGADREETRVVDSDYTQFALMLSRRHTSRLAVLRISLLGRSWLLPPGTLDQFICLGRAQGLSDDNIVFPDVTGNMVHLQACWAVGTGPAGMSLVDPRGAGPSVYPGSSAPACAQGSPGSWVPVLNPGSEPPPAAPGPLSWATSSHPGSPVPGHLLPPQVPCPGPPPPAPPAPGPLSRPTSSHPGSPVLGYLLPPQVPCPGPSPPSGSPVLGHLLPSPIPAHKELGLIPGGALDLSSLPWVAAPA
RUO
LCN12
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote